Learn More
3M™ Face Seals
Click to view available options
Color:
Black
White
Unit Size:
Each
Pack of 10
Pack of 5
Specifications
Specifications
| Color | White |
| Product Type | Face Seal |
| For Use With (Equipment) | Suspension headcovers and hoods |
| Quantity | 10/Pk. |