Learn More
Dräger™ Face Mask R52850 Full Panorama Nova RA
Product Code. 12342369
Change view
Click to view available options
Color:
Black
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
12342369 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12342369 Supplier Dräger™ Supplier No. R52850
  • ONLINE EXCLUSIVE PRICE
    4615.00 NOK
    4010.69 NOK / Pack of 1
    Save 604.31 NOK
    13% Off
Estimated Shipment: 29-01-2027
The Full Panorama Nova RA is ideal for applications involving hazardous materials, independence of the ambient air and where protection of the respiratory tract and eyes is needed. Designed for use with Rd40 filter range with it's RA connection. Filters must be purchased separately.
  • Full facemask
  • Stainless steel fittings
  • Rd40 standard thread connector
  • Mist-free visor
  • EPDM mask body, polycarbonate visor
  • Can be used at high and low temperatures
  • Comfortable fit - suitable for most face shapes
  • 180° wide angle lens and unrestricted field of vision
Specifications
Certifications/Compliance EN 136 class 3 (CE mark), DIN 58610, NIOSH, AS/NZS (depending on variant)
Color Black
Connection Type Rd 40 round thread x 1/7 in.
Product Type Full Facepiece Respirator
Material Mask body: EPDM, Visor: Polycarbonate