Learn More
FABP4 Mouse anti-Human, Unconjugated, Clone: 3E7E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67167-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The FABP4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FABP4. This antibody recognizes human, mouse, rat, pig antigen. The FABP4 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
FABP4 | |
Monoclonal | |
747 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, Immunohistochemistry | |
3E7E1 | |
Unconjugated | |
20 μL | |
2167 | |
IgG2b |