Learn More
FABP4 Mouse anti-Human, Unconjugated, Clone: 3E7E1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67167-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16879645
  • 4593.76 NOK / 150µL
Estimated Shipment: 24-05-2024
Description
The FABP4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FABP4. This antibody recognizes human, mouse, rat, pig antigen. The FABP4 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
FABP4
Monoclonal
747 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry
3E7E1
Unconjugated
150 μL
2167
IgG2b