missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP3/H-FABP Rabbit anti-Mouse, Rat, Clone: 9B9D7, Novus Biologicals™
Rabbit Monoclonal Antibody
1760.00 NOK - 4590.00 NOK
Specifications
| Antigen | FABP3/H-FABP |
|---|---|
| Clone | 9B9D7 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18307405
|
Novus Biologicals
NBP3-16508-20UL |
20 μg |
1760.00 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18320844
|
Novus Biologicals
NBP3-16508-100UL |
100 μg |
4590.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FABP3/H-FABP Monoclonal antibody specifically detects FABP3/H-FABP in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
| FABP3/H-FABP | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| FABP11, fatty acid binding protein 11, fatty acid binding protein 3, muscle and heart (mammary-derived growthinhibitor), Fatty acid-binding protein 3, Fatty acid-binding protein 3, muscle, fatty acid-binding protein, heart, Heart-type fatty acid-binding protein, H-FABPM-FABP, Mammary-derived growth inhibitor, MDGI, Muscle fatty acid-binding protein, O-FABP | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human FABP3/H-FABP (P05413). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 9B9D7 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Cholesterol Metabolism, Lipid and Metabolism | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 2170 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title