missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F2 Mouse anti-Human, Clone: 1E12A8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66509-1-IG-150UL
This item is not returnable.
View return policy
Description
Prothrombin or Coagulation factor II, is proteolytically cleaved to form thrombin in the first step of the coagulation cascade which ultimately results in the stemming of blood loss. F2 also plays a role in maintaining vascular integrity during development and postnatal life. Mutations in F2 leads to various forms of thrombosis and dysprothrombinemia.Specifications
| F2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P00734 | |
| F2 | |
| F2 Fusion Protein Ag18385 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1E12A8 | |
| Unconjugated | |
| F2 | |
| Coagulation factor II, F2, Prothrombin, PT | |
| Mouse | |
| Protein G | |
| RUO | |
| 2147 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction