Learn More
F1063-0967, MedChemExpress
Product Code. 16473398
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16473398
Brand: Medchem Express HY101510
MedChemExpress F1063-0967 is a Dual-specificity phosphatase 26 (DUSP26) inhibitor with an IC50 of 11.62 μM.
F1063-0967 is a Dual-specificity phosphatase 26 (DUSP26) inhibitor with an IC50 of 11.62 μM.
Specifications
Chemical Name or Material F1063-0967
CAS 613225-56-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H24N2O5S2
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (206.36 mM; Need ultrasonic)
SMILES OC1=C(C(O)=O)C=CC(NC(CCCCCN2C(S/C(C2=O)=C\C3=CC=C(C)C=C3)=S)=O)=C1
Molecular Weight (g/mol) 484.59
Formula Weight 484.59
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.