Learn More
Thermo Scientific™ F1-ClipTip™ Variable Volume Single Channel Pipettes
Lock tips firmly to the pipette with a complete seal with ClipTip technology, so they will not loosen or fall off. Lightweight handle design combined with minimal tip attachment and ejection forces.
Brand: Thermo Scientific™ 4641200N
3040.26 NOK valid until 2025-09-30
Use promo code "24314" to get your promotional price.
Description
Ensure a secure fit with ClipTip technology, which locks tips firmly to the pipette with a complete seal so they will not loosen or fall off. Lightweight handle design combined with minimal tip attachment and ejection forces will increase the conform in pipetting. Feel the difference.
Worry-free pipetting is finally here with the only tip that clips.
Sealed in security of ClipTip Technology. Feel the difference.
Eliminate the frustration of banging tips on your pipette
With a light touch, each tip is clipped securely on the pipette.
Reduce the risk of loose or leaking tips
When ClipTip pipette tips are on, they are sealed and will not loosen. The ClipTip pipetting system remains sealed until you decide to eject tips, even in the most comprehensive applications, including multiple tip touches and mixing cycles.
Enable consistent and reproducible pipetting from user to user
Higher-quality results and more efficient research. Learn more
Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of repetitive strain injuries
Secure Volume Lock
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort.
Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.
Improved small volume pipetting
New 0.1-2 ÎĽL model and improved versions 1-10 ÎĽL models allow optimized performance in your small volume applications.
Wide Selection of ClipTip Pipette Tips
Compatible sizes with F1-ClipTip pipettes: 12.5 Ext, 20, 50, 200, 300, 300 Ext and 1000ÎĽL
Life-time Warranty
With convenient online registration, and may vary by country.
Specifications
ClipTip 200 | |
Yellow | |
F1-ClipTip | |
10 to 100 ÎĽL | |
Yellow |
F1-ClipTip | |
1 | |
Variable Volume Pipette | |
0.2ÎĽL |
Promotions
-
Multiple Manufacturers
Sample Preparation, Filtration and Storage
Get up to 80% off some of our most popular products.Promo Code: 24124Expires: 2025-08-31 Redemption Only
-
Thermo Scientific ClipTip
,
Thermo Scientific Finnpipette
,
Mbp Molecular Bio Products
Huge Savings on Liquid Handling Essentials
Unlock Precision: Top Pipettes and Tips on Promotion!Promo Code: 24081Expires: 2025-12-31
-
Multiple Manufacturers
Genomics Powerhouse
Power your pipeline from PCR to sequencing.Promo Code: 24117Expires: 2025-08-24