Learn More
Thermo Scientific™ F1-ClipTip™ Multichannel Pipettes
Product Code. 15591935
Change view
Click to view available options
Volume (Metric):
1 to 10 μL
10 to 100 μL
30 to 300 μL
5 to 50 μL
Number of Channels:
8
12
Compatible Tips:
ClipTip 12.5 Ext
ClipTip 20
ClipTip 200
ClipTip 300
ClipTip 50
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Number of Channels Compatible Tips Unit Size
15591935 30 to 300 μL 8 ClipTip 300 Each
15561935 1 to 10 μL 8 ClipTip 20 Each
15571935 5 to 50 μL 8 ClipTip 50 Each
15581935 10 to 100 μL 8 ClipTip 200 Each
15501945 1 to 10 μL 12 ClipTip 20 Each
15511945 5 to 50 μL 12 ClipTip 50 Each
15521945 10 to 100 μL 12 ClipTip 200 Each
15531945 30 to 300 μL 12 ClipTip 300 Each
15936142 1 to 10 μL 8 ClipTip 12.5 Ext Each
15956142 1 to 10 μL 12 ClipTip 12.5 Ext Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 15591935 Supplier Thermo Scientific™ Supplier No. 4661140N
  • ONLINE EXCLUSIVE PRICE
    14343.37 NOK
    8813.74 NOK / Pack of 1
    Save 5529.63 NOK
    38% Off
View all promotions
In Stock
Add to basket
Add to basket
Bulk Quote Request
Lock tips firmly in place with ClipTip technology and ensure a complete seal on every channel with minimal tip attachment and ejection forces using Thermo Scientific™ Multichannel Pipettes.
Lock tips firmly in place with the Thermo Scientific™ F1-ClipTip™ Multichannel Pipettes, which use ClipTip™ tips that eject only when you intend them to. The ClipTip technology offers extremely low tip attachment and ejection forces and a complete seal on every channel. This can improve the reproducibility of your results. Enhanced volume adjustment allows for a silent, effortless and ergonomic user experience.
Worry-free pipetting is finally here with the only tip that clips.
Sealed in security of ClipTip Technology. Feel the difference.
Eliminate the frustration of banging tips on your pipette
With a light touch, each tip is clipped securely on the pipette.
Reduce the risk of loose or leaking tips
When ClipTip pipette tips are on, they are sealed and will not loosen. The ClipTip pipetting system remains sealed until you decide to eject tips, even in the most comprehensive applications, including multiple tip touches and mixing cycles.
Enable consistent and reproducible pipetting from user to user
Higher-quality results and more efficient research. Learn more
Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of repetitive strain injuries
Secure Volume Lock
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort.
Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.
Improved small volume pipetting
New 0.1-2 μL model and improved versions 1-10 μL models allow optimized performance in your small volume applications.
Wide Selection of ClipTip Pipette Tips
Compatible sizes with F1-ClipTip pipettes: 12.5 Ext, 20, 50, 200, 300, 300 Ext and 1000μL
Life-time Warranty
With convenient online registration, and may vary by country.
TRUSTED_SUSTAINABILITY
Specifications
Compatible Tips ClipTip 300
Description F1-ClipTip
Color Orange
Number of Channels 8
Product Line F1-ClipTip
Type Pipette
Volume (Metric) 30 to 300 μL
Increments 1μL