Learn More
Eye Wash Dispenser
Product Code. 12920485
Change view
Click to view available options
Capacity: 20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12920485 Capacity: 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12920485 Supplier Safety First Supplier No. E440
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The 1997 Approved Code of Practice states that Where tap water is not readily available for eye irrigation, at least 1L of sterile water or sterile normal saline 0.9% in sealed disposable containers should be provided.
Specifications
For Use With (Equipment) Eye Wash Pods
Height (Metric) 35 cm
Includes Integrated Mirror, Instructions
Mounting Wall