Learn More
MilliporeSigma™ Extrelut™ NT Packing
Product Code. 15875800
Change view
Click to view available options
Type:
Bulk Material
Refill Pack
For Use With (Equipment):
EXtrelut NT1, NT3 and NT20
EXtrelut NT20
Unit Size:
1kg
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) Unit Size
15875800 Refill Pack EXtrelut NT20 Pack of 50
15865800 Bulk Material EXtrelut NT1, NT3 and NT20 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15875800 Supplier MilliporeSigma™ Supplier No. 1150930001
 Restricted Item
Estimated Shipment: 04-01-2027
Use in the production of large extraction columns. MilliporeSigma EXtrelut™ NT Packing is made of pure Kieselguhr (CAS 68855-54-9, pH 10) and is available in a bulk pack for use with all Extrelut™ NT columns or pre-portioned refill packs specif
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) EXtrelut NT20
Type Refill Pack
compliance-icons
Product Identifier
  • EXtrelut NT Refill pack for column fillings EXtrelut NT 20
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Kieselguhr