Learn More
Lenz Laborglasintrumente Extractors Acc. to Soxhlet with Dimroth Condenser
Product Code. 10738841
Change view
Click to view available options
Capacity (Metric):
70 mL
100 mL
150 mL
300 mL
500 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10738841 300 mL Each
10382761 100 mL Each
10047850 150 mL Each
10352811 500 mL Each
10251511 70 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10738841 Supplier Lenz Laborglasintrumente Supplier No. 05360052
  • ONLINE EXCLUSIVE PRICE
    4785.00 NOK
    4263.39 NOK / Pack of 1
    Save 521.61 NOK
    10% Off
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • The method allows an effective separation with relatively small amounts of extracting solvents.
  • Extractor in compact and robust design, with flask connection NS 29/32
  • Made of DURAN™ tubing
  • Dimroth condenser with thread GL 14
  • Round bottom flask
Specifications
Capacity (Metric) 300 mL
Material Duran Glass
Standard Taper Joint NS 60/46