Learn More
Lenz Laborglasintrumente Extractors Acc. to Soxhlet with Allihn Condenser
Product Code. 10614012
Change view
Click to view available options
Capacity (Metric):
30 mL
70 mL
150 mL
200 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10614012 30 mL Each
10204631 200 mL Each
10234581 70 mL Each
10519581 150 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10614012 Supplier Lenz Laborglasintrumente Supplier No. 05365025
  • ONLINE EXCLUSIVE PRICE
    2490.00 NOK
    2148.18 NOK / Pack of 1
    Save 341.82 NOK
    13% Off
Estimated Shipment: 11-01-2027
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Extractor in compact and robust design
  • With flask connection
  • Made of DURAN™ tubing
  • Allihn condenser with thread GL 14
  • Round bottom flask
Specifications
Capacity (Metric) 30 mL
Material Duran Glass
Standard Taper Joint NS 29/32