Learn More
asecos™ Extraction Unit
Product Code. 10686644 Shop All asecos Products
Change view
Click to view available options
extraction air monitoring: no
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10686644 extraction air monitoring: no
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
For indoor use only
Product Code. 10686644 Supplier asecos™ Supplier No. 14218
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
For indoor use only
Extraction units/modules for installation on the cabinet or wall mounting – ready to plug-in, easy and quick installation
Extraction units/modules for installation on the cabinet or wall mounting – ready to plug-in, easy and quick installation
Safe capturing of escaping vapours and gases at the point where they are released or produced
  • Very low noise, ideal for direct installation at the workplace
  • With connection cable and power cord
  • Connecting fitting DN 75
  • ATEX compliant: C` II 3/-G Ex c llC T4
Specifications
Certifications/Compliance CE compliant: C ` II 3/- G Ex c IIC T4
Depth (Metric) Exterior 400 mm
Description Extraction Unit
Dimensions (D x W x H) 400 x 200 x 200 mm
For Use With (Application) Safe capturing of escaping vapours and gases at the point where they are released or produced
For Use With (Equipment) For tall cabinets
Height (Metric) Exterior 200 mm
Model HF.EA.8677.
Surface Coating Powder-coated plain
Weight (Metric) 10.1 kg
Width (Metric) Exterior 200 mm
Product Type Extraction Unit