missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Extracellular Vesicle Human CD9/CD63/CD81 Antibody Panel
Description
- The Extracellular Vesicle Human CD9/CD63/CD81 Antibody Panel is suitable for the detection of extracellular vesicles (EVs) derived from human cells
- It comprises primary antibodies that are immunoreactive toward human CD9, CD63, and CD81; these are proteins typically expressed on EVs and widely used as markers to analyze and isolate these cell-derived particles
- CD9, CD63, and CD81 belong to the tetraspanin family of membrane proteins, which possess four transmembrane domains and interact with diverse proteins on the cell surface to form multimolecular networks termed tetraspanin-enriched microdomains
- CD9, CD63, and CD81 proteins are expressed on the surface of many cells, including B cells, T cells, NK cells, monocytes, dendritic cells, thymocytes, endothelial cells, and fibroblasts, and are involved in modulating a variety of cellular processes including cell activation, adhesion, differentiation, and tumor invasion
- The antibodies provided in this panel have been reported for use in analyzing primary cells, cell lines, and EVs by ELISA, flow cytometry, immunocytochemistry, immunoprecipitation, and Western blotting
- They have been reported to cross-react with their cognate antigens in non-human primates, including baboons and rhesus and cynomolgus macaques
Specifications
Specifications
| Classification | Monoclonal |
| Host Species | Mouse |
| Quantity | 1 Kit |
| Cell Type | Other |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?