Learn More
Ismatec™ Extension Tubing, Tygon ST
Product Code. 10148551
Change view
Click to view available options
Tubing Size:
I.D. 0.19 mm
I.D. 0.38 mm
I.D. 0.57 mm
I.D. 0.89 mm
I.D. 1.14 mm
I.D. 1.65 mm
I.D. 1.85 mm
Unit Size:
10m
Each
Product Code. Tubing Size unitSize
10148551 I.D. 0.19 mm 10m
15753319 I.D. 0.38 mm Each
15868894 I.D. 0.57 mm 10m
15763319 I.D. 0.89 mm Each
10514033 I.D. 1.14 mm 10m
15773319 I.D. 1.65 mm Each
10027331 I.D. 1.85 mm 10m
7 options
Product Code. 10148551
Brand: Ismatec™ 0550210
Only null left
Add to basket
Add to basket
For use with 2- and 3-stop color-coded tubing.
  • Inexpensive, all-around tubing for general laboratory applications.
  • Transparent
  • Resistant to almost all inorganic chemicals
  • Tasteless
  • Smooth, polished inner wall
  • Low gas permeability
  • Non-aging and non-oxidizing
  • High dialectric constant
Specifications
Certifications/Compliance FDA 21 CFR175.300
Autoclavable Autoclavable
Diameter (Metric) Inner 0.19 mm
Material Tygon ST R-3603, R-3607
Overall Length (Metric) 10 m
For Use With (Application) General laboratory applications, 2- and 3-stop color-coded tubing.
Min. Operating Temperature (Metric) −50°C
Packaging Quantity 1 Reel
Resistance Properties Acids: Good, Alkaline solutions: Good, Solvents: Not rec., Pressure: Fair, Vacuum: Good, Viscous media: Excellent, Sterile media: Limited
Tubing Size I.D. 0.19 mm
Sterilization Method Autoclavable with steam, 30 minutes at 1 bar (15 psi) and 121°C (250°F); tubing will appear milky.Gas sterilization with Ethylene oxide.Not recommended for sterilization with radiation.
Color Transparent
Max. Operating Temperature (Metric) 74°C