Learn More
Buerkle™ Extension Rod
Product Code. 10175372 Shop All Bürkle Products
Change view
Click to view available options
Length (Metric):
500 mm
600 mm
1000 mm
Color:
Gray
Pure White
Silver
For Use With (Equipment):
ChemoSampler
Mole Drill Sampler
PTFE Scoop
PharmaPicker
PharmaPicker Pharma Sampler
Unit Size:
Each
Product Code. Length (Metric) Color For Use With (Equipment) unitSize
10175372 500 mm Silver PharmaPicker Pharma Sampler Each
10413522 600 mm Pure White PTFE Scoop Each
10033832 1000 mm Silver PharmaPicker Each
10661954 1000 mm Silver Mole Drill Sampler Each
10123152 1000 mm Gray ChemoSampler Each
5 options
Product Code. 10175372
Brand: Buerkle™ 53800506
Only null left
Add to basket
Add to basket
Buerkle™ Extension Rod makes it possible to take samples from various depths.
  • Seven different drill bits are available, each of which is designed for a particular soil type
  • For various applications
TRUSTED_SUSTAINABILITY
Specifications
Product Type Extension Rod
Material Stainless Steel (V4A)
Autoclavable Autoclavable
Description Equipment for PharmaPicker
Color Silver
Finish Electro-polished
For Use With (Equipment) PharmaPicker Pharma Sampler
Length (Metric) 500 mm