Learn More
Masterflex™ Extension Cable
Product Code. 12590887
Click to view available options
Size:
10 ft.
Unit Size:
Each
Product Code. 12590887
Brand: Masterflex™ 7709503
Only null left
Add to basket
Add to basket
Masterflex™ Extension Cable for increasing the length of Masterflex™ connection cables by three meters
  • Make the most of Masterflex™ pumps with connection cables, replacement parts, and other accessories
  • Choose accessories and replacement parts suitable for your Masterflex™ pump
  • Increase flexibility with extended cable lengths
Specifications
Size 10 ft.
Size (Metric) 3 m