Learn More
Cytiva Exposure Cassette for Unmounted Screen
Product Code. 15834182 Shop All Cytiva Products
Change view
Click to view available options
Dimensions (LxW):
20 x 25 cm
35 x 43 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (LxW) Unit Size
15834182 35 x 43 cm Each
15824182 20 x 25 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15834182 Supplier Cytiva Supplier No. 29175524
 Restricted Item
Estimated Shipment: 10-12-2026
Protects the screen from light during exposure to the sample.
  • For use with unmounted screens (not included).
  • Protects your screen from light during exposure.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Light exposure
For Use With (Equipment) Unmounted Screen
Dimensions (LxW) 35 x 43 cm