missing translation for 'onlineSavingsMsg'
Learn More

EXOSC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15062-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16894044

  • 3828.53 NOK / 150µL
Estimated Shipment: 09-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

EXOSC3 is component of the exosome 3′->5′ exoribonuclease complex and is required for the 3′ processing of the 7S pre-RNA to the mature 5.8S rRNA.
Specifications

Specifications

EXOSC3
Polyclonal
Unconjugated
EXOSC3
CGI 102, EXOSC3, exosome component 3, hRrp 40, hRrp40p, p10, RP11 3J10.8, RRP40, Rrp40p
Rabbit
Antigen Affinity Chromatography
RUO
313243, 51010, 66362
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q7TQK4, Q9NQT5
Exosc3
EXOSC3 Fusion Protein Ag7065
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers