missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXOSC10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16731-1-AP-20UL
This item is not returnable.
View return policy
Description
Human DNA-binding protein (dbpA) is a member of a Y-box binding protein family containing a cold shock domain. The increased expression of Y box binding proteins in somatic cells is associated with cell proliferation and transformation.Specifications
| EXOSC10 | |
| Polyclonal | |
| Unconjugated | |
| EXOSC10 | |
| Autoantigen PM/Scl 2, EXOSC10, exosome component 10, p2, p3, p4, PM Scl, PM/Scl 100, PMSCL, PMSCL2, RRP6, Rrp6p | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 50912, 5394 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P56960, Q01780 | |
| Exosc10 | |
| EXOSC10 Fusion Protein Ag10189 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction