Learn More
EXOSC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12585-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16880364
  • 4593.76 NOK / 150µL
Estimated Shipment: 22-05-2024
Description
This gene encodes a core component of the exosome. The mammalian exosome is required for rapid degradation of AU rich element-containing RNAs but not for poly(A) shortening. The association of this protein with the exosome is mediated by protein-protein interactions with ribosomal RNA-processing protein 42 and ribosomal RNA-processing protein 46.
Specifications
Specifications
EXOSC1
Polyclonal
Unconjugated
EXOSC1
CGI 108, CSL4, Csl4p, EXOSC1, exosome component 1, hCsl4p, p13, SKI4, Ski4p
Rabbit
Antigen Affinity Chromatography
RUO
51013, 66583, 679140
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9DAA6, Q9Y3B2
EXOSC1
EXOSC1 Fusion Protein Ag3286
150 μL
Primary
Human, Mouse, Rat