missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXOSC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12585-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a core component of the exosome. The mammalian exosome is required for rapid degradation of AU rich element-containing RNAs but not for poly(A) shortening. The association of this protein with the exosome is mediated by protein-protein interactions with ribosomal RNA-processing protein 42 and ribosomal RNA-processing protein 46.Specifications
EXOSC1 | |
Polyclonal | |
Unconjugated | |
EXOSC1 | |
CGI 108, CSL4, Csl4p, EXOSC1, exosome component 1, hCsl4p, p13, SKI4, Ski4p | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51013, 66583, 679140 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9DAA6, Q9Y3B2 | |
EXOSC1 | |
EXOSC1 Fusion Protein Ag3286 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |