Learn More
Applied Biosystems™ ExoSAP-IT™ Express PCR Product Cleanup Reagent
Product Code. 15573677
Change view
Click to view available options
No. of Reactions:
100
2000
480 reactions, 8-tube strip
500
5000
Unit Size:
100 reactions
2000 reactions
480 reactions
500 reactions
5000 reactions
Product Code. No. of Reactions unitSize
15573677 2000 2000 reactions
15543677 500 500 reactions
15563677 100 100 reactions
15553677 5000 5000 reactions
15533677 480 reactions, 8-tube strip 480 reactions
5 options
Product Code. 15573677
Brand: Applied Biosystems™ 75001.4X.1.ML
Edge
Removes excess primers and dNTPs. Does not interfere with downstream applications.
  • Mixture of two hydrolytic enzymes, a modified Exonuclease I and Shrimp Alkaline Phosphatase (SAP), in a specially formulated buffer
  • One-tube, one-step PCR cleanup
  • Five-minute protocol
  • Single tube, 8-tube strip, and 96-well plate formats offer flexibility from manual pipetting to automated workflows
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C
No. of Reactions 2000