missing translation for 'onlineSavingsMsg'
Learn More

EXOC6B Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55440-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16846905

  • 3828.53 NOK / 150µL
Estimated Shipment: 17-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Component of the exocyst complex involved in the docking of exocytic vesicles with fusion sites on the plasma membrane.
Specifications

Specifications

EXOC6B
Polyclonal
Unconjugated
EXOC6B
EXOC6B, exocyst complex component 6B, KIAA0919, SEC15 like protein 2, SEC15B, SEC15L2
Rabbit
Antigen Affinity Chromatography
RUO
23233, 75914
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
0.34 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A6H5Z3, Q9Y2D4
Exoc6b
Peptide
150 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers