missing translation for 'onlineSavingsMsg'
Learn More

EXOC6B Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16856905 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16856905 20 μL 20µL
16846905 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16856905

Brand: Proteintech 554401AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Component of the exocyst complex involved in the docking of exocytic vesicles with fusion sites on the plasma membrane.

Specifications

Antigen EXOC6B
Applications Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EXOC6B
Gene Accession No. A6H5Z3, Q9Y2D4
Gene Alias EXOC6B, exocyst complex component 6B, KIAA0919, SEC15 like protein 2, SEC15B, SEC15L2
Gene Symbols Exoc6b
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23233, 75914
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.