missing translation for 'onlineSavingsMsg'
Learn More

EXOC6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16820604 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16820604 150 μL 150µL
16830604 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16820604

Brand: Proteintech 127231AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The product of this gene belongs to the SEC15 family. It is highly similar to the protein encoded by Saccharomyces cerevisiae SEC15 gene. This protein is essential for vesicular traffic from the Golgi apparatus to the cell surface in yeast. It is one of the components of a multiprotein complex required for exocytosis. Alternatively spliced transcript variants encoding different isoforms have been identified.

Specifications

Antigen EXOC6
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EXOC6
Gene Accession No. O54923, Q8R313, Q8TAG9
Gene Alias EXOC6, EXOC6A, exocyst complex component 6, FLJ1125, SEC15, SEC15 like protein 1, SEC15A, SEC15L, SEC15L1, SEC15L3, Sec15p
Gene Symbols EXOC6
Host Species Rabbit
Immunogen EXOC6 Fusion Protein Ag3418
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107371, 50556, 54536
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.