Learn More
Justrite Exhaust Fan Lithium Battery Cabinet, No Sparks
Product Code. 17722388
Change view
Click to view available options
:
Exhaust Fan Lithium Battery Cabinet, No Sparks; Dimensions (L x W): 29 x 29 cm; Material: Steel
Product Code.
17722388 Exhaust Fan Lithium Battery Cabinet, No Sparks; Dimensions (L x W): 29 x 29 cm; Material: Steel
1 options
Product Code. 17722388
Brand: Justrite JCBDM125BP
Exhaust fan with filter holding box for high cabinet with 1 or 2 doors.
  • Exhaust fan with filter holding box for high cabinet with 1 or 2 doors
  • Outlet diameter: 125 mm (filter excluded)
Specifications
Length (Metric) 29 cm
Width (Metric) 29 cm
Material Steel
Product Type Exhaust Fan Cabinet