Learn More
Thermo Scientific Chemicals Exemestane
Product Code. 16691822 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16691822 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16691822 Supplier Thermo Scientific Chemicals Supplier No. 463160010
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Molecular Weight (g/mol) 296.41
Product Type Exemestane
Quantity 1 g