Learn More
Epredia™ Excelsior™ AS Tissue Processor Accessories
Product Code. 15594280 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Air System Duct Adapter Kit
Basket Carrying Container
Charcoal Filter
Downdraft Extraction Duct Adapter
Monitor Clamp Kit
Organized Basket
Potassium Permanganate Filter
Storage Bottle
Waste Wax Drawer Kit
Unit Size:
Each
Pack of 6
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15594280 Organized Basket Each
12638156 Downdraft Extraction Duct Adapter Each
12648156 Air System Duct Adapter Kit Each
12683156 Storage Bottle Pack of 6
12678156 Waste Wax Drawer Kit Each
12688156 Monitor Clamp Kit Each
12608346 Basket Carrying Container Each
13470487 Charcoal Filter Pack of 6
12764856 Potassium Permanganate Filter Pack of 6
12658156 Organized Basket Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 15594280 Supplier Epredia™ Supplier No. A82310038
  • ONLINE EXCLUSIVE PRICE
    10350.00 NOK
    9005.83 NOK / Pack of 1
    Save 1344.17 NOK
    12% Off
In Stock
Maintain the Epredia™ Excelsior™ AS Tissue Processor with Epredia™ Excelsior AS Tissue Processor Accessories.
Product Features:
  • Microwaveable design allowing pre-processing of tissue
  • Organized capacity of 300 cassettes in six basket-halves
Ordering Alerts:
Basket halves only match with each other but can be mixed with 222 cassette baskets or SecureSette Excelsior ES baskets
Includes:
Six 50-cassette basket halves and user guide and loading instructions
Compatible with:
Excelsior ES Tissue Processor
TRUSTED_SUSTAINABILITY
Specifications
Product Type Organized Basket
For Use With (Equipment) Excelsior AS Tissue Processor
Basket Capacity 300 Cassettes
Content And Storage Ambient