Learn More
Epredia™ Excelsior™ AS Tissue Processor Accessories
Product Code. 12658156 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Air System Duct Adapter Kit
Basket Carrying Container
Downdraft Extrctn Duct Adapter
Monitor Clamp Kit
Organized Basket, 222 cassette
Organized Basket, 300 cassette
Potassium Permanganate Filter
Storage Bottle
Vapor Filter (Charcoal)
Waste Wax Drawer Kit
Unit Size:
Each
Pack of 6
Product Code. Product Type unitSize
12658156 Organized Basket, 222 cassette Pack of 6
12638156 Downdraft Extrctn Duct Adapter Each
12648156 Air System Duct Adapter Kit Each
12683156 Storage Bottle Pack of 6
12678156 Waste Wax Drawer Kit Each
12688156 Monitor Clamp Kit Each
12608346 Basket Carrying Container Each
13470487 Vapor Filter (Charcoal) Pack of 6
12764856 Potassium Permanganate Filter Pack of 6
15594280 Organized Basket, 300 cassette Each
10 options
Product Code. 12658156
Brand: Epredia™ A78410025
Maintain the Epredia™ Excelsior™ AS Tissue Processor with Epredia™ Excelsior AS Tissue Processor Accessories.
For use with Shandon Excelsior ES Tissue Processor
TRUSTED_SUSTAINABILITY
Specifications
Product Type Organized Basket, 222 cassette
For Use With (Equipment) Excelsior AS Tissue Processor
Basket Capacity 222 Cassettes