Learn More
Isofield™ Exa 230 Validated Sterile Earloop Cleanroom Facemask with Clip - Grade A/B Compatible
Click to view available options
:
Size: Wide
Description
- Validated sterile earloop cleanroom facemask with clip
- 230 mm length with malleable nose strip
- 3-layer non-woven configuration
- Highly efficient bacterial and particle filtration
- Gamma irradiation, minimum 25 kGy
- Sterility Assurance Level 10-6
- Particle Shedding (Helmke Drum) Category I Individually packed pouches printed with IPA resistant ink to maintain traceability
Specifications
Specifications
| Product Type | Cleanroom Face Mask |
| Color | White |
| Fastening Type | Earloops |
| Material | Polyethylene, Polypropylene |
| Size | Wide |
| Sterility | Sterile |
| Style | Pleated |