Learn More
CleanSpace™ EX Power System
Shop All CleanSpace Technology Products
Click to view available options
:
Power Unit
Description
- Intrinsically safe
- Light weight and comfortable
- High visibility yellow casing for easy recognition
- AirSensit System™ for mask pressure control and breath responsive airflow
- One-button system and standby and auto-start/stop mode for quick don/doffing
- Easy to use support harness
- Altitude compensated filter blocked detection
- Long operating time: up to 8 hours and quick battery recharging: < 2hours
- Easy and accurate flow capability check
Specifications
Specifications
| Product Type | PAPR Assembly, No Headtop |
| Model | PAF-0060 |
Mask sold separately