missing translation for 'onlineSavingsMsg'
Learn More

EWS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55191-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16896675

  • 3828.53 NOK / 150µL
Estimated Shipment: 17-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is specific to EWS. A 90 kDa band also could be detected.

This gene encodes a multifunctional protein that is involved in various cellular processes, including gene expression, cell signaling, and RNA processing and transport. The protein includes an N-terminal transcriptional activation domain and a C-terminal RNA-binding domain. Chromosomal translocations between this gene and various genes encoding transcription factors result in the production of chimeric proteins that are involved in tumorigenesis. These chimeric proteins usually consist of the N-terminal transcriptional activation domain of this protein fused to the C-terminal DNA-binding domain of the transcription factor protein. Mutations in this gene, specifically a ttranslocation, are known to cause Ewing sarcoma as well as neuroectodermal and various other tumors. Alternative splicing of this gene results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 1 and 14.
Specifications

Specifications

EWS
Polyclonal
Unconjugated
EWSR1
EWS, EWS oncogene, EWSR1, RNA binding protein EWS
Rabbit
Antigen Affinity Chromatography
RUO
14030, 2130, 289752
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.57 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q01844, Q61545
Ewsr1
Peptide
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers