Learn More
EVOS™ EVOS™ Vessel Holder, one 100-mm Petri dish
Click to view available options
Quantity:
1 Holder
Unit Size:
Each
Description
Broad range of holders helps maximize the flexibility of the EVOS and Micromaster™ inverted microscopes.
Specifications
Specifications
| Type | Vessel Holder |
| Product Line | EVOS |
| For Use With (Equipment) | EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System |
| Format | one 100-mm Petri dish |
| Holds | 1 100 mm Petri Dish |
| Quantity | 1 Holder |