Learn More
Invitrogen™ EVOS™ Light Cube, YFP
Product Code. 11957285
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. 11957285
Brand: Invitrogen™ AMEP4654
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The revolutionary design of the EVOS™ light cubes combines a high intensity, long lifetime light source (LED), condensing optics, dichroic mirror and filters into a compact, adjustable, easy-to-install accessory for fluorescence detection on the EVOS™ imaging systems and the Countess II FL.
  • Ideal for use with EVOS FL Color Imaging System, EVOS FL Auto Imaging System or EVOS FL Imaging Systems
  • Each cube contains an LED, condensing optics, and hard-coated filters
  • Continuous illumination enables consistent results 50,000+ hour bulb lifetime that affords lower operating costs
  • Adjustable light intensity that helps reduce photobleaching User changeable and automatically recognized by the system
TRUSTED_SUSTAINABILITY
Specifications
Type YFP Fluorescent Light Cube
Product Line EVOS
For Use With (Equipment) Countess™ II FL Automated Cell Counter, EVOS™ M5000 Imaging System, EVOS™ M7000 Imaging System
Includes LED, Condensing Optics, Hard-coated Filters
Wavelength Ex: 500/24 Em: 542/27
Quantity 1 Ea.