Learn More
EVOS™ EVOS™ 100X Oil Objective, achromat, coverslip-corrected
Description
- Coverslip-corrected objective that is optimized for imaging through #1.5 coverslips
- Offers optical performance from visible light to near infrared light
- Meets needs across spectrum of magnifications and optical specifications
Specifications
Specifications
| Type | Objective Lens |
| Product Line | EVOS |
| For Use With (Equipment) | EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System |
| Includes | Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates |
| Focus Distance | 0.17 mm |
| Lens Type | Achromat |
| Magnification | 100X |
| Numerical Aperture | 1.25 |
| Quantity | 1 each |