Learn More
Thermo Scientific™ eVol™ XR Dispensing System Battery
Order the Thermo Scientific™ eVol™ Dispensing System Battery when it needs replacing.
Brand:  Thermo Scientific™ 66002-029
Additional Details : Weight : 0.20000kg
Product Code. 13456786
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Order the Thermo Scientific™ eVol™ Dispensing System Battery when it needs replacing. The eVol Dispensing System is a unique, hand-held digital dispenser accurately and reproducibly performs a wide variety of liquid handling procedures. The easy-to-use device couples an analytical syringe to a programmable electronic controller, enabling highly reproducible dispensing for all users.
The eVol Dispensing System is ideal for laboratories that need to repeatedly dispense nonaqueous liquids, offering greater accuracy and reproducibility for applications such as:
  • Routine dispensing of volatile solvents, hazardous, corrosive or viscous chemicals
  • Preparation of calibration standards
  • Addition of internal standards
The system unifies two precision devices: a digitally controlled electronic drive, and a Thermo Scientific™ XCHANGE™ enabled analytical syringe.
Repeatable Performance for Higher Confidence in Your Results
  • Can be calibrated easily by a user to ensure validity of results
  • Intuitive, easy-to-use interface with touch wheel and full-color screen
  • Complies with GLP and GMP guidelines
Exceptional Versatility and Functionality
  • Integrated XCHANGE coupling allows syringes to be quickly and easily changed, preventing possible cross-contamination
  • Addition of Thermo Scientific™ MEPS™ (Micro Extraction by Packed Sorbent) technology to an XCHANGE syringe enables automated sample preparation by SPE in a hand-held system