Learn More
Thermo Scientific™ eVol™ XR Dispensing System
Product Code. 10752826 Shop All Thermo Scientific Products
Product Code. 10752826
Brand: Thermo Scientific™ 66002020
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Eliminate dispensing variability with this unique, hand-held digital dispenser that accurately and reproducibly performs a wide variety of liquid handling procedures.
Conventional pipetting can sometimes lead to errors and rework, depending on which technician is doing the work. Now there's a way to eliminate dispensing variability and improve efficiency. The Thermo Scientific™ eVol™ Dispensing System is a unique, hand-held digital dispenser that accurately and reproducibly performs a wide variety of liquid handling procedures. The easy-to-use device couples an analytical syringe to a programmable electronic controller, enabling highly reproducible dispensing for all users.
The eVol Dispensing System is ideal for laboratories that need to repeatedly dispense nonaqueous liquids, offering greater accuracy and reproducibility for applications such as:
  • Routine dispensing of volatile solvents, hazardous, corrosive or viscous chemicals
  • Preparation of calibration standards
  • Addition of internal standards
The system unifies two precision devices: a digitally controlled electronic drive, and a Thermo Scientific™ XCHANGE™ enabled analytical syringe.
Repeatable Performance for Higher Confidence in Your Results
  • Can be calibrated easily by a user to ensure validity of results
  • Intuitive, easy-to-use interface with touch wheel and full-color screen
  • Complies with GLP and GMP guidelines
Exceptional Versatility and Functionality
  • Integrated XCHANGE coupling allows syringes to be quickly and easily changed, preventing possible cross-contamination
  • Addition of Thermo Scientific™ MEPS™ (Micro Extraction by Packed Sorbent) technology to an XCHANGE syringe enables automated sample preparation by SPE in a hand-held system
TRUSTED_SUSTAINABILITY
Specifications
Description Sample Dispensing System
For Use With (Application) Routine Dispensing of Volatile Solvents, Hazardous, Corrosive or Viscous Chemicals
Product Type Electronic Dispensing System
Product Line HyperSep