Learn More
Emma Safety Footwear Evoke S2 Shoe
Anatomically designed for a superior fit, the Evoke is a shoe that's been designed for long days in and out of truck cabs, building sites, warehouses - wherever your work takes you.
Brand: Emma Safety Footwear MM74054748
Additional Details : Weight : 1.01000kg
Description
- S2-Safety shoe for wet working conditions
- Metal Free
- The energy returning Evoke. Anatomically designed for a superior fit, the Evoke is a shoe that's been designed for long days in and out of truck cabs, building sites, warehouses - wherever your work takes you.
- The shoe's tongue has been ingeniously designed to prevent moisture and dirt entering.
- Comfortable, supportive with energy returning soles, the Evoke's a low version of the Everon.
- Industries:
- Logistics & transport
Specifications
Laces | |
Hydro-Tec Antibacterial | |
48 | |
ISO 20345:2011 | |
S2 |
Black, Gray | |
Nubuck | |
No | |
Standard width |