Learn More
Emma Safety Footwear Evoke S2 Shoe
Anatomically designed for a superior fit, the Evoke is a shoe that's been designed for long days in and out of truck cabs, building sites, warehouses - wherever your work takes you.
Brand:  Emma Safety Footwear MM74054744
Additional Details : Weight : 1.01000kg
 View more versions of this product
Product Code. 17532546
  • 1197.80 NOK / 1 pair
Estimated Shipment: 31-05-2024
Description
  • S2-Safety shoe for wet working conditions
  • Metal Free
  • The energy returning Evoke. Anatomically designed for a superior fit, the Evoke is a shoe that's been designed for long days in and out of truck cabs, building sites, warehouses - wherever your work takes you.
  • The shoe's tongue has been ingeniously designed to prevent moisture and dirt entering.
  • Comfortable, supportive with energy returning soles, the Evoke's a low version of the Everon.
  • Industries:
    • Logistics & transport
Specifications
Specifications
Laces
Hydro-Tec Antibacterial
44
ISO 20345:2011
S2
Black, Gray
Nubuck
No
Standard width