missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ EVI2A (Human) Recombinant Protein
Human EVI2A full-length ORF ( AAH35572, 25 a.a. - 236 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00002123-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: SPGTKANYTRLWANSTSSWDSVIQNKTGRNQNENINTNPITPEVDYKGNSTNMPETSHIVALTSKSEQELYIPSVVSNSPSTVQSIENTSKSHGEIFKKDVCAENNNNMAMLICLIIIAVLFLICTFLFLSTVVLANKVSSLRRSKQVGKRQPRSNGDFLASGLWPAESDTWKRTKQLTGPNLVMQSTGVLTATRERKDEEGTEKLTNKQIG
Specifications
AAH35572 | |
ecotropic viral integration site 2A | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EVI2A | |
GST |
2123 | |
Wheat germ expression system | |
25 μg | |
EVDA, EVI2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |