Learn More
Lenz Laborglasinstrumente™ Evaporating Flask with Conical Ground Joint
Product Code. 10509271
Change view
Click to view available options
Capacity: 2000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10509271 Capacity: 2000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10509271 Supplier Lenz Laborglasinstrumente™ Supplier No. 03042973
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Especially for use in rotary evaporators. Lenz Laborglasinstrumente™ Evaporating Flasks with Conical Ground Joints can be used to make solutions or to hold, contain, collect or measure chemicals and are manufactured from durable DURAN™ glass.
  • With center neck and conical joint
  • Pear-shaped with inscription label
  • Available in a range of capacities
Specifications
Capacity (Metric) 2000 mL
Material Borosilicate Glass, Duran Glass
For Use With (Equipment) Rotary Evaporators
Neck Style Center
Diameter (Metric) Outer 158 mm