Learn More
Fisherbrand™ Sealing Mat for 96-Well Deep Square Nonsterile Microplates
Click to view available options
Product Type:
96 Well Square Sealing Cap
Unit Size:
Pack of 50
Description
- For friction sealing of plates during transport or storage
- Cap mats are an economic alternative to thermal or adhesive seals
- Cap mats can be sterilized and reused
- Constructed of high quality ethylene-vinyl acetate (EVA)
- Rnase/Dnase free
Specifications
Specifications
| Color | White |
| Material | EVA |
| Sterility | Non-sterile |
| No. of Wells | 96 |
| No. per Pack | 50 |
| Product Type | 96 Well Square Sealing Cap |
| Well Shape | Square |
| Plate Compatibility | 96-well Square Deepwell Plate |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.