Learn More
Eutech™ pH Buffers and Solutions
Description
The measure of a liquid's acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.
Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes.
Features:
- Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Specifications
Specifications
| Color | Undesignated |
| Description | pH buffer kit, clear buffer solutions each. |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 4, 7, 10 |
| Physical Form | Liquid |
| Quantity | 480 mL |
| Solution Type | pH Buffer |
| Packaging | Kit with 480mL bottles |