Learn More
Eutech™ Conductivity Calibration Sachets
Description
Get quick, accurate, and convenient conductivity measurements with NIST_traceable Eutech™ Conductivity Calibration Sachets. Simply insert electrode into sachet pack, calibrate, rinse and discard sachet – no separate container needed.
Features:
- Conductivity Standards Sachet -20 ml
- NIST Traceable
Specifications
Specifications
| Chemical Name or Material | KCl Conductivity Sachets |
| Conductivity | 2764 μS/cm |
| Solution Type | Calibration Solution |
| Type | Conductivity Standard |
| Industry Type | Field, School, Laboratory Use |