Learn More
Brady™ European AC Adapter for Printer
Description
- Charge your printer for a full day of battery.
Specifications
Specifications
| Product Type | AC Adapter |
| Color | Black |
| For Use With (Equipment) | BMP21-LAB Label Printer, BMP21-PLUS Label Printer, Brady M210 Label Printer, Brady M210-LAB Label Printer |
| Description | AC Adaptor, Europe 220 V |
| Format | AC Adapter |
| Quantity | 1 Ea. |
| Voltage | 220 V |
| Alternate Part Number | BMP21-AC EUR |