Learn More
Euro Protection™ Eurmask Uno
Product Code. 15684137
Click to view available options
:
COMPATIBILITY: 22130, 22140, 22150, 22160, 22170, 22210
Product Code. 15684137
Brand: Euro Protection™ 22130
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Euro Protection™ Eurmask Uno is a half mask for a single threaded cartridge.
  • Half mask for 1 threaded cartridge
  • Body: TPE thermoplastic rubber
  • Harness: Fabric and rubber elastic strap
  • Valves: Silicone
  • Adjustable straps
Specifications
Certifications/Compliance EN 140 :1998
Material TPE Thermoplastic rubber, silicone
For Use With (Equipment) COMPATIBILITY: 22130, 22140, 22150, 22160, 22170, 22210
Quantity 1/Pk., 16Pk./Cs.
Weight (Metric) 115 g