Learn More
Eugenol, Sigma-Aldrich™
Specifications
Specifications
| CAS | 97-53-0 |
| Melting Point | -9.2°C |
| Density | 1.07 g/cm3 at 20°C |
| Boiling Point | 252°C/1013 hPa |
| Molecular Formula | 2-(CH3O)-4-(CH2=CHCH2)C6H3OH |
| MDL Number | MFCD00008654 |
| Quantity | 100 mL |
| Synonym | Eugenol, 4-Allyl-2-methoxyphenol, Eugenol |
| Molecular Weight (g/mol) | 164.2 |
| Grade | Research |