Learn More
Etoricoxib, MedChemExpress
Product Code. 16466736
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466736 100 mg 100mg
16452557 50 mg 50mg
16416458 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16466736 Supplier Medchem Express Supplier No. HY15321
MedChemExpress Etoricoxib (MK-0663) is a non steroidal anti-inflammatory agent, acting as a selective and orally active COX-2 inhibitor, with IC50s of 1.1 μM and 116 μM for COX-2 and COX-1 in human whole blood.
Etoricoxib (MK-0663) is a non steroidal anti-inflammatory agent, acting as a selective and orally active COX-2 inhibitor, with IC50s of 1.1 μM and 116 μM for COX-2 and COX-1 in human whole blood.
Specifications
Chemical Name or Material Etoricoxib
CAS 202409-33-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H15ClN2O2S
Quantity 100 mg
Synonym MK-0663 L-791456
Solubility Information DMSO : 100 mg/mL (278.68 mM; Need ultrasonic)
SMILES O=S(C1=CC=C(C2=CC(Cl)=CN=C2C3=CC=C(C)N=C3)C=C1)(C)=O
Molecular Weight (g/mol) 358.84
Formula Weight 358.84
Percent Purity 99.1%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation